missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Phospho-DVL2 (Ser143) Recombinant Rabbit Monoclonal Antibody (K01_1Q43)

Product Code. 30825115
Change view
Click to view available options
Quantity:
50 μL
100 μL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity
30825115 50 μL
30824635 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30825115 Supplier Invitrogen™ Supplier No. MA564869

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Recombinant Monoclonal Antibody

Phospho-DVL2 (Ser143) Recombinant Monoclonal Antibody for Western Blot

Disheveled DVL2 is a member of the dishevelled (dsh) protein family. The vertebrate dsh proteins have approximately 40% amino acid sequence similarity with Drosophila dsh. This gene encodes a 90-kD protein that undergoes posttranslational phosphorylation to form a 95-kD cytoplasmic protein, which may play a role in the signal transduction pathway mediated by multiple Wnt proteins. The mechanisms of dishevelled function in Wnt signaling are likely to be conserved among metazoans.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Phospho-DVL2 (Ser143)
Applications Western Blot
Classification Recombinant Monoclonal
Clone K01_1Q43
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation 50mM tris glycine with 40% glycerol, 150mM NaCl, 0.05% BSA and 0.01% sodium azide; pH 7.4
Gene DVL2
Gene Accession No. O14641, Q60838
Gene Alias dishevelled 2; dishevelled 2 (homologous to Drosophila dsh); dishevelled 2, dsh homolog; dishevelled 2, dsh homolog (Drosophila); dishevelled segment polarity protein 2; dishevelled, dsh homolog 2; dishevelled-2; DSH homolog 2; Dvl2; Dvl-2; LOW QUALITY PROTEIN: segment polarity protein dishevelled homolog DVL-2; sb:eu471; Segment polarity protein dishevelled homolog DVL-2; wu:fc05d12; wu:fo71e09; wu:fp54a02; zgc:55372
Gene Symbols DVL2
Host Species Rabbit
Immunogen A synthetic phosphopeptide corresponding to residues surrounding Ser143 of human Dishevelled 2.
Purification Method Protein A/G
Quantity 50 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 13543, 1856
Target Species Human, Mouse
Content And Storage Maintain refrigerated at 2-8°C for up to 1 month. For long term storage store at -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.