missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Phospho-ELK1 (Ser383) Polyclonal Antibody

Phospho-ELK1 (Ser383) Polyclonal Antibody
Brand: Invitrogen PA597336
This item is not returnable.
View return policy
Description
Phospho-ELK1 (Ser383) Polyclonal Antibody for Western Blot, ICC/IF
Specifications
| Phospho-ELK1 (Ser383) | |
| Polyclonal | |
| Unconjugated | |
| ELK1 | |
| ELK 1; ELK1; Elk-1; ELK1, ETS transcription factor; ELK1, member of ETS oncogene family; ETS domain-containing protein Elk-1; ETS transcription factor ELK1; ETS-like gene 1; Oncogene Elk1; tyrosine kinase (ELK1) oncogene | |
| Rabbit | |
| Affinity Chromatography | |
| RUO | |
| 2002 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| ELISA, Immunohistochemistry (Paraffin), Western Blot | |
| 1.3 mg/mL | |
| PBS with 50% glycerol and 0.09% sodium azide; pH 7.3 | |
| P19419 | |
| ELK1 | |
| A synthetic phosphorylated peptide around S383 of human ELK1 (NP_0052202). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction