missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Phospho-EPHA2 (Tyr588) Rabbit Polyclonal Antibody, Proteintech
Rabbit Polyclonal Antibody
€ 135.00 - € 360.00
Specifications
| Antigen | EPHA2 (Tyr588) |
|---|---|
| Concentration | 500 μg/mL |
| Applications | Western Blot, ELISA |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
30236253
|
Proteintech
30263-1-AP-20UL |
20 μL |
€ 135.00
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
30237094
|
Proteintech
30263-1-AP-100UL |
100 μL |
€ 360.00
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
The Phospho-EPHA2 (Tyr588) antibody from Proteintech is a rabbit polyclonal antibody generated with peptide of human Phospho-EPHA2 (Tyr588). This antibody recognizes human antigen. The Phospho-EPHA2 (Tyr588) antibody has been validated for the following applications: WB, ELISA analysis.Specifications
| EPHA2 (Tyr588) | |
| Western Blot, ELISA | |
| Unconjugated | |
| Rabbit | |
| Human | |
| ECK | |
| IgG | |
| Antigen affinity purification | |
| Polyclonal Antibody |
| 500 μg/mL | |
| Polyclonal | |
| Liquid | |
| RUO | |
| 1969 | |
| Peptide | |
| Primary | |
| Store at -20°C. Stable for one year after shipment. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title