missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Phospho-FGFR4 (Tyr642) Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA5105531
This item is not returnable.
View return policy
Description
Antibody detects endogenous levels of FGFR4 only when phosphorylated at Tyr642.
FGFR4 is a member of the FGFR family of receptor tyrosine kinases. This family is known to regulate a host of cellular functions including angiogenesis, mitogenesis, osteogenesis, myogenesis, carcinogenesis, cellular differentiation, and tissue repair after injury. The FGFR family has also been implicated in a number of diseases including cancer, rheumatoid arthritis, and diabetic retinopathy. FGFR4 is required for normal down-regulation of the expression of CYP7A1, the rate-limiting enzyme in bile acid synthesis, in response to FGF19. FGFR4 plays a role in postnatal lung development and may be involved in the development of skeletal muscle cell lineages.
Specifications
| Phospho-FGFR4 (Tyr642) | |
| Polyclonal | |
| Unconjugated | |
| Fgfr4 | |
| CD334; CTLA-2-beta protein; FGF Receptor; FGF Receptor 4; FGFR; Fgfr4; FGFR-4; Fibroblast growth factor receptor 4; fibroblast growth factor receptor 4 16 minus form; hydroxyaryl-protein kinase; JTK2; MGC20292; Mpk-11; protein-tyrosine kinase; protein-tyrosine kinase receptor MPK-11; TKF; tyrosine kinase related to fibroblast growth factor receptor; tyrosylprotein kinase | |
| Rabbit | |
| Sequential chromatography | |
| RUO | |
| 14186, 2264, 25114 | |
| -20°C | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
| 1 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
| P22455, Q03142, Q498D6 | |
| Fgfr4 | |
| A synthesized peptide derived from human FGFR4(Accession P22455), corresponding to amino acid residues around phosphorylated Tyr642. | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction