missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Phospho-FLT3 (Tyr599) Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Invitrogen PA538273
This item is not returnable.
View return policy
Description
This gene encodes a class III receptor tyrosine kinase that regulates hematopoiesis. The receptor consists of an extracellular domain composed of five immunoglobulin-like domains, one transmembrane region, and a cytoplasmic kinase domain split into two parts by a kinase-insert domain. The receptor is activated by binding of the fms-related tyrosine kinase 3 ligand to the extracellular domain, which induces homodimer formation in the plasma membrane leading to autophosphorylation of the receptor. The activated receptor kinase subsequently phosphorylates and activates multiple cytoplasmic effector molecules in pathways involved in apoptosis, proliferation, and differentiation of hematopoietic cells in bone marrow. Mutations that result in the constitutive activation of this receptor result in acute myeloid leukemia and acute lymphoblastic leukemia.
Specifications
| Phospho-FLT3 (Tyr599) | |
| Polyclonal | |
| Unconjugated | |
| FLT3 | |
| FLT3, Fms-like tyrosine kinase 3, receptor-type tyrosine-protein kinase FLT3, CD135, FL cytokine receptor, fetal liver kinase 2, stem cell tyrosine kinase 1 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 14255, 2322 | |
| -20°C | |
| Liquid |
| Western Blot | |
| 1 mg/mL | |
| Dulbecco′s PBS with 150mM NaCl, 50% glycerol and 0.02% sodium azide; pH 7.4 | |
| P36888, Q00342 | |
| FLT3 | |
| A synthetic phosphopeptide derived from human FLT3 around the phosphorylation site of Tyr599 (Y-E-YP-D-L) | |
| 100 μg | |
| Primary | |
| Human, Mouse, Non-human Primate | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction