missing translation for 'onlineSavingsMsg'
Läs mer
Läs mer
Invitrogen™ Phospho-FOXO1/FOXO3 (Ser207, Ser212) Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ 441230G
Denna artikel kan inte returneras.
Se returpolicy
Beskrivning
Phospho-FOXO1/FOXO3 (Ser207, Ser212) Polyclonal Antibody for Western Blot
a transcription factor of the forkhead family. A central regulator of metabolism in several cell types. May play an important role in myogenic growth and differentiation, and in the regulation of lipolysis in adipocytes by controlling the expression of adipose triglyceride lipase (ATGL), the rate-limiting lipolytic enzyme. Translocation of this gene with PAX3 has been associated with alveolar rhabdomyosarcoma. Contains 1 fork-head domain.
Specifikationer
| Phospho-FOXO1/FOXO3 (Ser207, Ser212) | |
| Polyclonal | |
| Dulbecco's PBS with 1mg/mL BSA and 0.05% sodium azide; pH 7.3 | |
| O43524, Q12778, Q9R1E0, Q9WVH4 | |
| FOXO1, FOXO3 | |
| The antiserum was produced against a chemically synthesized phosphopeptide derived from the region of human FOXO3 that contains serine 207. | |
| 100 μL | |
| Primary | |
| Human, Mouse | |
| Antibody | |
| IgG |
| Western Blot | |
| Unconjugated | |
| FOXO3 | |
| 1110048B16Rik; 2010203A17Rik; AF6q21; AF6q21 protein; AFX; AFX1; Afxh; AI876417; C76856; FKH1; Fkhr; Fkhr1; Fkhr2; Fkhrl1; FKHRL1P2; forkhead box O1; forkhead box O1a; forkhead box O1A (rhabdomyosarcoma); forkhead box O3; forkhead box O3A; forkhead box protein O1; Forkhead box protein O1A; forkhead box protein O3; forkhead homolog (rhabdomyosarcoma) like 1; Forkhead in rhabdomyosarcoma; Forkhead in rhabdomyosarcoma-like 1; forkhead protein 1; forkhead protein FKHR2; forkhead, Drosophila, homolog of, in rhabdomyosarcoma; forkhead, Drosophila, homolog of, in rhabdomyosarcoma-like 1; FOXO1; Foxo1a; FOXO2; FOXO3; Foxo3a; FOXO4; MGC120490; MLLT7 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 2308, 2309, 56458, 56484 | |
| -20°C | |
| Liquid |
Korrigering av produktinnehåll
Din input är viktig för oss. Fyll i det här formuläret för att ge feedback relaterad till innehållet på denna produkt.
Produkttitel
Hittar du en möjlighet till förbättring?Dela en innehållskorrigering