missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Phospho-GAB1 (Tyr627) Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ 44568G
This item is not returnable.
View return policy
Description
Phospho-GAB1 (Tyr627) Polyclonal Antibody for Western Blot
Gab1 is a 115 kDa multiple docking protein that plays an essential role in cellular growth, transformation and apoptosis. Gab1 can be phosphorylated by multiple receptor tyrosine kinase (RTKs), including: insulin receptor (IR), platelet-derived growth factor receptor beta] (PDGFRbeta]), hepatocyte growth factor/scatter factor receptor (HGFR/SFR or c-Met), and epidermal growth factor receptor (EGF), as well as in response to cell-cell adhesion. Gab1 is tyrosine phosphorylated on at least 16 sites, some of which serve as binding sites for phosphoatidylinositol 3-kinase (PI3K), Grb2, PLC gamma 1, Nck, and SHP2. Phosphorylation of Gab1 on tyrosines 627 and 659 is critical for its binding to SHP2, and for activation of the ERK/MAPK pathway in response to EGF.
Specifications
| Phospho-GAB1 (Tyr627) | |
| Polyclonal | |
| Dulbecco's PBS with 1mg/mL BSA and 0.05% sodium azide; pH 7.3 | |
| Q13480, Q9QYY0 | |
| GAB1 | |
| The antiserum was produced against a chemically synthesized phosphopeptide derived from the region of human Gab1 that contains tyrosine 627. The sequence is conserved in mouse. | |
| 100 μL | |
| Primary | |
| Human, Mouse | |
| Antibody | |
| IgG |
| Western Blot | |
| Unconjugated | |
| GAB1 | |
| AA408973; AW107238; EGK_16125; GAB1; GRB2 associated binding protein 1; GRB2-associated binder 1; GRB2-associated binding protein 1; GRB2-associated binding protein 1 long isoform; GRB2-associated-binding protein 1; Growth factor receptor bound protein 2-associated protein 1; OTTHUMP00000220263 | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 14388, 2549 | |
| -20°C | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction