missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Phospho-GAB2 (Ser159) Polyclonal Antibody

Phospho-GAB2 (Ser159) Polyclonal Antibody
Brand: Invitrogen PA5105241
This item is not returnable.
View return policy
Description
Phospho-GAB2 (Ser159) Polyclonal Antibody for Western Blot
Specifications
| Phospho-GAB2 (Ser159) | |
| Polyclonal | |
| Unconjugated | |
| GAB2 | |
| AI463667; D130058I17Rik; GAB2; Grb2 associated binder 2; GRB2 associated binding protein 2; GRB2-associated binder 2; GRB2-associated binding protein 2; GRB2-associated-binding protein 2; GRB3-3; Growth factor receptor bound protein 2-associated protein 2; KIAA0571; p97; PH domain-containing adaptor molecule p97; pp100 | |
| Rabbit | |
| Sequential Chromatography | |
| RUO | |
| 14389, 84477, 9846 | |
| -20°C | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot | |
| 1 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
| Q9EQH1, Q9UQC2, Q9Z1S8 | |
| GAB2 | |
| A synthesized peptide derived from human GAB2(Accession Q9UQC2), corresponding to amino acid residues around phosphorylated Ser159. | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction