missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Phospho-gamma-H2AX (Ser139) Recombinant Rabbit Monoclonal Antibody (BLR053F)
GREENER_CHOICE

Rabbit Recombinant Monoclonal Antibody

€ 156.00 - € 477.00

Specifications

Antigen Phospho-gamma-H2AX (Ser139)
Clone BLR053F
Concentration 100 μg/mL
Content And Storage 4°C
Applications Flow Cytometry, Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot, Immunocytochemistry
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
17755669
GREENER_CHOICE
View Documents
Invitrogen™
MA544299
100 μL
€ 477.00
100µL
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
17765669
View Documents
Invitrogen™
MA544300
10 μL
€ 156.00
10µL
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
Description

Description

Epitope retrieval with citrate buffer pH6.0 is recommended for FFPE cell sections.

H2AX is a variant of the H2A protein family and is a component of the histone octomer in nucleosomes. H2AX is phosphorylated by kinases such as ataxia telangiectasia mutated (ATM) and ATM-Rad3-related (ATR) in the PI3K pathway. Gamma-H2AX is the first step in recruiting and localizing DNA repair proteins. DSBs can be induced by mechanisms such as ionizing radiation or cytotoxic agents and subsequently, gamma-H2AX foci quickly form. These foci represent the DSBs in a 1:1 manner and can be used as a biomarker for damage. The visualization of gamma-H2AX by can allow the assessment of DNA damage, related DNA damage proteins and DNA repair. Gamma-H2AX has applications in the detection of genomic damage caused by cytotoxic chemical agents and environmental and physical damage.
Specifications

Specifications

Phospho-gamma-H2AX (Ser139)
100 μg/mL
Flow Cytometry, Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot, Immunocytochemistry
Unconjugated
Rabbit
RUO
BBS with 0.1% BSA and 0.09% sodium azide; pH 8.2
P16104
3014
Peptide surrounding phosphorylated serine 140 of human histone H2AX.
Antibody
BLR053F
4°C
Recombinant Monoclonal
Liquid
IgG
Human
H2AX
gamma H2A X; gamma H2A.X; gamma-H2A.X; gh2ax
H2AX
Primary
purified
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.