missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Phospho-GATA1 (Ser310) Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA5104243
This item is not returnable.
View return policy
Description
Antibody detects endogenous levels of GATA1 only when phosphorylated at Serine 310.
Transcriptional activator or repressor which probably serves as a general switch factor for erythroid development. It binds to DNA sites with the consensus sequence 5'-[AT]GATA[AG]-3' within regulatory regions of globin genes and of other genes expressed in erythroid cells.
Specifications
| Phospho-GATA1 (Ser310) | |
| Polyclonal | |
| Unconjugated | |
| Gata1 | |
| eryf1; Erythroid transcription factor; erythroid transcription factor 1; GATA binding protein 1; GATA binding protein 1 (globin transcription factor 1); GATA1; GATA-1; GATA-binding factor 1; GATA-binding protein 1; GATA-binding protein 1 (globin transcription factor 1); GF1; Gf-1; globin transcription factor 1; HGNC:4170; NFE1; NF-E1; NF-E1 DNA-binding protein; nuclear factor, erythroid 1; transcription factor GATA1; XLANP; XLTDA; XLTT | |
| Rabbit | |
| sequential chromatography | |
| RUO | |
| 14460, 25172, 2623 | |
| -20°C | |
| Liquid |
| Western Blot, Immunocytochemistry | |
| 1 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
| P15976, P17679, P43429 | |
| Gata1 | |
| A synthesized peptide derived from human GATA1(Accession P15976), corresponding to amino acid residues around phosphorylated Ser310. | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat, Monkey | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction