missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Phospho-GSK3 alpha/beta (Tyr216, Tyr279) Recombinant Rabbit Monoclonal Antibody (4A5)

Rabbit Recombinant Monoclonal Antibody

Brand:  Invitrogen™ MA560620

Product Code. 30534422

  • € 482.00 / 100µL

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

Phospho-GSK3 alpha/beta (Tyr216, Tyr279) Recombinant Monoclonal Antibody for IHC (P), ELISA

Glycogen synthase kinase-3 (GSK3) is a protein kinase that was originally identified as a regulator of glycogen synthase, a key enzyme in glycogen metabolism. Since then, it has been shown to be involved in the regulation of a diverse array of cellular functions, including protein synthesis, cell proliferation, cell differentiation, microtubule assembly/disassembly, and apoptosis. GSK3's substrate specificity is unique in that phosphorylation of the substrate only occurs if a phosphoserine or phosphotyrosine is present - four residues C-terminal to the site of GSK phosphorylation. There exists two isoforms of GSK3, GSK3 alpha and GSK3 beta, and they are strictly regulated via phosphorylation. Phosphorylation of GSK3 beta on Ser9 (Ser21 in GSK3alpha) by protein kinase B (PKB) causes its inactivation and is the primary mechanism responsible for growth factor inhibition of this kinase. Activation of GSK3 beta is dependent upon the phosphorylation of Tyr216 (Tyr279 in GSK3 alpha). Upon activation, it has been shown to phosphorylate a number of different cellular proteins, including p53, c-Myc, c-Jun, heat shock factor-1 (HSF-1), and cyclin D1. GSK3 beta also has been shown to phosphorylate aberrant sites on the microtubule associated protein tau, which is critical for the progression of Alzheimer's disease.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

Phospho-GSK3 alpha/beta (Tyr216, Tyr279)
Recombinant Monoclonal
0.6 mg/mL
PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
P49840, P49841
GSK3A, GSK3B
A synthesized peptide derived from human Phospho-GSK3A/GSK3B (Y216 + Y279).
100 μL
Primary
Human
Antibody
IgG
ELISA, Immunohistochemistry (Paraffin)
4A5
Unconjugated
GSK3B
2700086H06Rik; 7330414F15Rik; 8430431H08Rik; C86142; FA; factor A; glycogen synthase kinase 3 alpha; glycogen synthase kinase 3 beta; glycogen synthase kinase-3 alpha; Glycogen synthase kinase-3 beta; GSK3; GSK-3; GSK-3 alpha; GSK-3 beta; Gsk3a; gsk-3-alpha; Gsk3b; GSK-3beta; GSK3beta isoform; serine/threonine-protein kinase GSK3A; Serine/threonine-protein kinase GSK3B
Rabbit
Affinity chromatography
RUO
2931, 2932
-20°C or -80°C if preferred
Liquid
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.