missing translation for 'onlineSavingsMsg'
Learn More

HBP1 Polyclonal Antibody
GREENER_CHOICE

Product Code. 16673455
Change view
Click to view available options
Quantity:
100 μL
Unit Size:
100µL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16673455 100 μL 100µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 16673455 Supplier Invitrogen Supplier No. PA599207

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Phospho-HBP1 (Ser402) Polyclonal Antibody

Phospho-HBP1 (Ser402) Polyclonal Antibody for Western Blot, ICC/IF

TRUSTED_SUSTAINABILITY

Specifications

Antigen HBP1
Applications Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
Gene HBP1
Gene Accession No. O60381, Q62661, Q8R316
Gene Alias 1700058O05Rik; BE629963; C330012F01Rik; C86454; Hbp1; High mobility group box transcription factor 1; high mobility group protein 1; HMG box transcription factor 1; HMG box-containing protein 1; Hmgb1; HMG-box containing protein 1; HMG-box transcription factor 1
Gene Symbols HBP1
Host Species Rabbit
Immunogen A synthesized peptide derived from human HBP1(Accession O60381), corresponding to amino acid residues K355-S405.
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 26959, 27080, 73389
Target Species Human, Mouse, Rat
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.