missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Phospho-HICE1 (Ser19, Ser20) Polyclonal Antibody
Description
Recommended positive controls: HeLa, K562, Blocking peptide (GTX70011-PEP). Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
Hice1 contributes to mitotic spindle assembly, maintenance of centrosome integrity and completion of cytokinesis as part of the HAUS augmin-like complex.
Specifications
Specifications
| Antigen | Phospho-HICE1 (Ser19, Ser20) |
| Applications | ELISA, Western Blot, Immunocytochemistry |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Formulation | PBS with no preservative |
| Gene | HAUS8 |
| Gene Accession No. | Q9BT25 |
| Gene Alias | 2410004L22Rik; 4HAUS augmin-like complex, subunit 8; DGT4; HAUS augmin like complex subunit 8; HAUS augmin-like complex subunit 8; HAUS augmin-like complex, subunit 8; HAUS8; HEC1/NDC80 interacting, centrosome associated 1; HEC1/NDC80-interacting centrosome-associated protein 1; Hec1-interacting and centrosome-associated 1; Hice1; MGC20533; NY SAR 48; NYSAR48; NY-SAR-48; Sarcoma antigen NY-SAR-48; sarcoma antigen NY-SAR-48 homolog |
| Gene Symbols | HAUS8 |
| Host Species | Rabbit |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?