missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Phospho-HP1 gamma (Ser83) Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA517210
This item is not returnable.
View return policy
Description
It is not recommended to aliquot this antibody. This antibody is not cross-reactive with HP1-alpha or HP1-beta proteins.
At the nuclear envelope, the nuclear lamina and heterochromatin are adjacent to the inner nuclear membrane. The protein encoded by this gene binds DNA and is a component of heterochromatin. This protein also can bind lamin B receptor, an integral membrane protein found in the inner nuclear membrane. The dual binding functions of the encoded protein may explain the association of heterochromatin with the inner nuclear membrane.
Specifications
| Phospho-HP1 gamma (Ser83) | |
| Polyclonal | |
| Unconjugated | |
| Cbx3 | |
| CBX3; chromobox 3; chromobox homolog 3; chromobox homolog 3 (HP1 gamma homolog, Drosophila); chromobox protein homolog 3; chromobox-like protein 3; HECH; heterochromatin protein 1 gamma; heterochromatin protein 1 homolog gamma; heterochromatin protein HP1 gamma; heterochromatin-like protein 1; HGNC:1553; HP1 gamma; HP1 gamma homolog; HP1g; HP1-GAMMA; HP1Hs-gamma; HPy; M32; Modifier 2 protein; OTTHUMP00000202463 | |
| Rabbit | |
| Affinity chromatography | |
| RUO | |
| 100054158, 11335, 12417, 297093, 539788 | |
| -20°C | |
| Liquid |
| Immunoprecipitation, Western Blot, Immunocytochemistry | |
| 484 μg/mL | |
| 0.01M HEPES with 0.15M NaCl, 100 μg/mL BSA, 50% glycerol and no preservative, pH 7.5 | |
| P23198, Q13185 | |
| Cbx3 | |
| Synthetic phosphopeptide corresponding to amino acids surrounding pSer83 of human HP1 gamma. | |
| 100 μL | |
| Primary | |
| Bovine, Drosophila, Horse, Human, Mouse, Monkey, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction