missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Phospho-HSL (Ser565) Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA517487
This item is not returnable.
View return policy
Description
It is not recommended to aliquot this antibody. This antibody is not cross-reactive with Ser563 phosphorylated HSL.
Hormone-sensitive lipase (HSL) is an intracellular neutral lipase found in a variety of tissues, but primarily in adipose and steroidogenic tissues. It hydrolyzes triglycerides and cholesteryl esters. It is the major enzyme responsible for the mobilization of free fatty acids from adopise tissue.
Specifications
| Phospho-HSL (Ser565) | |
| Polyclonal | |
| Unconjugated | |
| LIPE | |
| 4933403G17Rik; AOMS4; EC 3.1.1.79; FPLD6; hormone - sensitive lipase testicular isoform; hormone-sensitive lipase; hormone-sensitive lipase testicular isoform; HSL; HSL antibody; LHS; lipase E; lipase E, hormone sensitive type; lipase, hormone sensitive; lipase, hormone-sensitive; LIPE; LIPS | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 16890, 3991 | |
| -20°C | |
| Liquid |
| Western Blot, Immunocytochemistry | |
| 5 μg/mL | |
| 0.01M HEPES with 100μg/mL BSA, 50% glycerol, 0.15M NaCl and no preservative; pH 7.5 | |
| P54310, Q05469 | |
| LIPE | |
| Synthetic phosphopeptide corresponding to residues surrounding pSer554 of human HSL (Ser565 of rat HSL). | |
| 100 μL | |
| Primary | |
| Human, Mouse | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction