missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Phospho-HSP27 (Ser78) Rabbit Polyclonal Antibody, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 28900-1-AP-100UL
This item is not returnable.
View return policy
Description
The Phospho-HSP27 (Ser78) antibody from Proteintech is a rabbit polyclonal antibody generated with peptide of human Phospho-HSP27 (Ser78). This antibody recognizes human, mouse, rat antigen. The Phospho-HSP27 (Ser78) antibody has been validated for the following applications: WB, ELISA analysis.Specifications
| HSP27 (Ser78) | |
| Polyclonal | |
| Unconjugated | |
| Rabbit | |
| Antigen affinity purification | |
| RUO | |
| 3315 | |
| Store at -20°C. Stable for one year after shipment. | |
| Polyclonal Antibody | |
| IgG |
| Western Blot, ELISA | |
| 165 μg/mL | |
| CMT2F; DKFZp586P1322; HMN2B; HS.76067; HSP27; HSP28; Hsp25; SRP27 | |
| Peptide | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| 27-30 kDa | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction