missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Phospho-IGF1R beta (Tyr1165, Tyr1166) Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA537602
This item is not returnable.
View return policy
Description
A suggested positive control for Western blot is 293 cells; suggested positive control for ICC/IF is MCF-7 cells.
IGF-1R (Insulin-like Growth Factor-1 Receptor) consists of alpha- and beta-subunits, which are disulfide-linked in a beta-alpha-alpha-beta configuration in the mature receptor. The alpha-subunit is completely extracellular, while the beta-subunit spans the membrane and the intracellular portion has intrinsic tyrosine kinase activity.
Specifications
| Phospho-IGF1R beta (Tyr1165, Tyr1166) | |
| Polyclonal | |
| Unconjugated | |
| IGF1R | |
| A330103N21Rik; CD221; D930020L01; hyft; IGF1R; IGF-1R; IGF-I receptor; IGFIR; IGFIRC; IGFR; Igfr1; insulin like growth factor 1 receptor; insulin-like growth factor 1 receptor; Insulin-like growth factor 1 receptor alpha chain; Insulin-like growth factor 1 receptor beta chain; Insulin-like growth factor I receptor; JTK13; line 186; soluble IGF1R variant 1; soluble IGF1R variant 2 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 16001, 25718, 3480 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot | |
| 1 mg/mL | |
| PBS with 0.5% BSA, 50% glycerol and 0.02% sodium azide; pH 7.4 | |
| P08069, P24062, Q60751 | |
| IGF1R | |
| Peptide sequence around phosphorylation site of tyrosine 1165/tyrosine 1166 (T-D-Y(p)-Y(p)-R-K) derived from Human IGF-1R. | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction