missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Phospho-IGF1R beta (Tyr1316) Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA578017
This item is not returnable.
View return policy
Description
Positive Control: IGF-1 treated MCF-7 cell Predicted Reactivity: Xenopus laevis (100%), Pig (100%), Rhesus Monkey (100%), Bovine (100%) Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
IGF-1R (Insulin-like Growth Factor-1 Receptor) consists of alpha- and beta-subunits, which are disulfide-linked in a beta-alpha-alpha-beta configuration in the mature receptor. The alpha-subunit is completely extracellular, while the beta-subunit spans the membrane and the intracellular portion has intrinsic tyrosine kinase activity.
Specifications
| Phospho-IGF1R beta (Tyr1316) | |
| Polyclonal | |
| Unconjugated | |
| IGF1R | |
| A330103N21Rik; CD221; D930020L01; hyft; IGF1R; IGF-1R; IGF-I receptor; IGFIR; IGFIRC; IGFR; Igfr1; insulin like growth factor 1 receptor; insulin-like growth factor 1 receptor; Insulin-like growth factor 1 receptor alpha chain; Insulin-like growth factor 1 receptor beta chain; Insulin-like growth factor I receptor; JTK13; line 186; soluble IGF1R variant 1; soluble IGF1R variant 2 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 3480 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot | |
| 1.83 mg/mL | |
| PBS with 1% BSA, 20% glycerol and 0.025% ProClin 300; pH 7 | |
| P08069 | |
| IGF1R | |
| Carrier-protein conjugated synthetic peptide surrounding phospho Tyr1316 of human IGF1R. The exact sequence is proprietary. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction