missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Phospho-IkB epsilon (Ser22) Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Invitrogen PA538070
This item is not returnable.
View return policy
Description
Inhibits NF-kappa-B by complexing with and trapping it in the cytoplasm. Inhibits DNA-binding of NF-kappa-B p50-p65 and p50-c-Rel complexes.
Specifications
| Phospho-IkB epsilon (Ser22) | |
| Polyclonal | |
| Unconjugated | |
| NFKBIE | |
| I-kappa-B-epsilon, IKBE, IkappaBepsilon, NF-kappa-BIE, NF-kappaB inhibitor epsilon, NFKBIE | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 18037, 316241, 4794 | |
| -20°C | |
| Liquid |
| Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot | |
| 1 mg/mL | |
| Dulbecco′s PBS with 150mM NaCl, 50% glycerol and 0.02% sodium azide; pH 7.4 | |
| O00221, O54910 | |
| NFKBIE | |
| A synthetic phosphopeptide derived from human IkB-epsilon around the phosphorylation site of Ser22 (I-E-SP-L-R) | |
| 100 μg | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction