missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Phospho-INSR (Tyr1361) Polyclonal Antibody
GREENER_CHOICE

Product Code. 16621205
Change view
Click to view available options
Quantity:
100 μL
Unit Size:
100µL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16621205 100 μL 100µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 16621205 Supplier Invitrogen™ Supplier No. PA5104754

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

Antibody detects endogenous levels of IR only when phosphorylated at Tyrosine 1361.

INSR is a receptor tyrosine kinase. Binding of insulin to the extracellular domain of INSR stimulates glucose uptake. INSR and IGF-1 receptors share major structural and functional similarity. The earliest cellular response to insulin stimulation is autophosphorylation of tyrosine in INSR. In humans, the INSR gene is located on chromosome 19. Defects in INSR are the cause of various insulin resistance syndromes and IGF-1R defects may also cause some forms of growth retardation.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Phospho-INSR (Tyr1361)
Applications Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
Gene INSR
Gene Accession No. P06213, P15127, P15208
Gene Alias 4932439J01Rik; CD 220; CD220; CD220 antigen; CD220 beta; CD221; D630014A15Rik; HHF 5; HHF5; IGF-I receptor; INSR; Insulin receptor; insulin receptor preproprotein; Insulin receptor subunit alpha; Insulin receptor subunit beta; IR; IR 1; IR beta; IR1; IR-1; IR-A; IR-B
Gene Symbols INSR
Host Species Rabbit
Immunogen A synthesized peptide derived from human INSR(Accession P06213), corresponding to amino acid residues around phosphorylated Tyr1361.
Purification Method sequential chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 16337, 24954, 3643
Target Species Human, Mouse, Rat
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.