missing translation for 'onlineSavingsMsg'
Learn More

Phospho-IR/IGF1R (Tyr1158) Polyclonal Antibody, Alexa Fluor™ 488, Invitrogen™

Product Code. 17114679
Change view
Click to view available options
Quantity:
100 μL
Unit Size:
100µL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
17114679 100 μL 100µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 17114679 Supplier Invitrogen Supplier No. 44802A1

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

This antibody is predicted to react with mouse and rat.

Insulin and Insulin-like Growth Factor receptors are transmembrane tyrosine kinase receptors that play critical roles in development, cell growth and metabolism. Insulin/Insulin-like growth factor-1 binding to the extracellular domain of IR and IGFR leads to autophosphorylation of the receptor and activation of the intrinsic tyrosine kinase activity, which allows appropriate substrates to be phosphorylated. Phosphorylation sites that are unique to each receptor presumably play a key role in these signaling differences. The catalytic loops within the tyrosine kinase domains contain a three tyrosine motif corresponding to Tyr1158, 1162 and 1163 (for the IR) and Tyr1131, 1135 and 1136 (for the IGF1R). Autophosphorylation of the tyrosine sites within the activation loop proceeds in a processive manner initiating at the second tyrosine (1162 or 1135), followed by phosphorylation at tyrosines 1158 (IGF-1R tyrosine 1131), 1163 (IGF-1R tyrosine 1136), resulting in the full activation of IR and IGF1R.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Phospho-IR/IGF1R (Tyr1158)
Applications Immunocytochemistry, Immunofluorescence
Classification Polyclonal
Conjugate Alexa Fluor 488
Formulation Dulbecco's PBS with 0.2% BSA and 0.05% sodium azide, pH 7.3
Gene IGF1R
Gene Accession No. P06213, P08069
Gene Alias 4932439J01Rik, A330103N21Rik, CD220, CD221, D630014A15Rik, D930020L01, HHF5, hyft, IGF1 Receptor, IGF-1 receptor, IGF1 Receptor beta, Igf1r, IGF-1R, IGF1R beta, IGF-I receptor, IGF-I Receptor beta, IGFIR, IGFIRC, IGFR, Igfr1, Insr, insulin like growth factor 1 receptor, insulin receptor, insulin receptor precursor, insulin receptor preproprotein, Insulin receptor subunit alpha, Insulin receptor subunit beta, Insulin-like growth factor 1 receptor, Insulin-like growth factor 1 receptor alpha chain, Insulin-like growth factor 1 receptor beta chain, insulin-like growth factor 1 receptor beta subunit, insulin-like growth factor I receptor, insulin-like growth factor receptor 1, insulin-like growth factor receptor type 1, insulin-like growth factor-I receptor, insulin-like growth factor-I receptor precursor, IR, IR-A, IR-B, JTK13, line 186, soluble IGF1R variant 1, soluble IGF1R variant 2
Gene Symbols IGF1R, Insr
Host Species Rabbit
Immunogen Synthetic phosphopeptide derived from the region of IR/IGF1R that contains tyrosine 1158 of the human insulin receptor (IR) (tyrosine 1131 for IGF1R)
Purification Method Size-exclusion chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 3480, 3643
Target Species Human
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.