missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Phospho-IRE1 alpha (Ser724) Recombinant Rabbit Monoclonal Antibody (K01_2K97)
GREENER_CHOICE

Product Code. 30824852
Change view
Click to view available options
Quantity:
50 μL
100 μL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity
30824852 100 μL
30828823 50 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30824852 Supplier Invitrogen™ Supplier No. MA563976

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Recombinant Monoclonal Antibody

Phospho-IRE1 alpha (Ser724) Recombinant Monoclonal Antibody for Western Blot

ERN1 serine/threonine kinase or endoplasmic reticulum to nucleus signaling 1 is the ER to nucleus signaling 1 protein, a human homologue of the yeast Ire1 gene product which possesses intrinsic kinase activity and an endoribonuclease activity and it is important in endoplasmic reticulum-based stress signals. ERN1 controls IRE1 proteolysis in mammalian cells. ERN1 has a critical function in extraembryonic cells that is essential for fetal viability. Specifically, IRE1 acts as the sensor of unfolded proteins in the ER that initiates transmittance of the unfolded protein signal from the ER to the nucleus by splicing XBP1 mRNA converting it into a potent unfolded-protein response transcriptional activator. Null mutants are viable, but IRE1 is essential for viability under stress conditions that cause unfolded proteins to accumulate in the ER. Depletion of IRE1p through the expression of a dominant negative form of IRE1p has no effect on transfected cells, but cell death via apoptosis occurs under stress conditions that cause unfolded proteins to accumulate in the ER.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Phospho-IRE1 alpha (Ser724)
Applications Western Blot
Classification Recombinant Monoclonal
Clone K01_2K97
Concentration 0.6 mg/mL
Conjugate Unconjugated
Formulation 50mM tris glycine with 0.05% BSA, 150mM NaCl, 40% glycerol and 0.01% sodium azide; pH 7.4
Gene ERN1
Gene Accession No. O75460
Gene Alias 9030414B18Rik; AI225830; C85377; endoplasmic reticulum (ER) to nucleus signalling 1; endoplasmic reticulum to nucleus signaling 1; endoplasmic reticulum to nucleus signalling 1; endoplasmic reticulum-to-nucleus signaling 1; Endoribonuclease; ER to nucleus signalling 1; ERN1; FLJ30999; hIRE1p; inositol-requiring 1; inositol-requiring 1 alpha; inositol-requiring enzyme 1; inositol-requiring protein 1; IRE1; IRE1a; IRE1a antibody; Ire1alpha; Ire1-alpha; Ire1p; IRE1p antibody; MGC163277; MGC163279; protein kinase/endoribonuclease; RGD1559716; Serine/threonine-protein kinase; Serine/threonine-protein kinase/endoribonuclease IRE1; serine/threonine-protein kinase/endoribonuclease IRE1-like
Gene Symbols ERN1
Host Species Rabbit
Immunogen A synthetic phosphopeptide corresponding to residues surrounding Ser724 of human IRE1.
Purification Method Protein A/G
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 2081
Target Species Human
Content And Storage Maintain refrigerated at 2-8°C for up to 1 month. For long term storage store at -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.