Learn More
Phospho-IRS1 (Tyr1179) Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand:  Invitrogen 44822G
Product Code. 10558163
  • € 516.00 / 100µL
Description
Phospho-IRS1 (Tyr1179) Polyclonal antibody specifically detects Phospho-IRS1 (Tyr1179) in Human samples. It is validated for Western Blot.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Phospho-IRS1 (Tyr1179)
Polyclonal
Dulbecco's PBS with 1mg/mL BSA, 50% glycerol and 0.05% sodium azide; pH 7.3
P35568
IRS1
The antiserum was produced against a chemically synthesized phosphopeptide derived from the region of IRS-1 that contains tyrosine 1179. The sequence is conserved among multiple species including human, mouse, rat and chicken.
100 μL
Primary
Human
Antibody
IgG
Western Blot
Unconjugated
IRS1
G972R; HIRS-1; Insulin receptor substrate 1; IRS1; IRS-1; IRS1IRM; pp185
Rabbit
Antigen affinity chromatography
RUO
3667
-20°C