Learn More
Phospho-IRS1 (Tyr1179) Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Invitrogen 44822G
This item is not returnable.
View return policy
Description
Phospho-IRS1 (Tyr1179) Polyclonal antibody specifically detects Phospho-IRS1 (Tyr1179) in Human samples. It is validated for Western Blot.
Specifications
| Phospho-IRS1 (Tyr1179) | |
| Polyclonal | |
| Dulbecco's PBS with 1mg/mL BSA, 50% glycerol and 0.05% sodium azide; pH 7.3 | |
| P35568 | |
| IRS1 | |
| The antiserum was produced against a chemically synthesized phosphopeptide derived from the region of IRS-1 that contains tyrosine 1179. The sequence is conserved among multiple species including human, mouse, rat and chicken. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
| Western Blot | |
| Unconjugated | |
| IRS1 | |
| G972R; HIRS-1; Insulin receptor substrate 1; IRS1; IRS-1; IRS1IRM; pp185 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 3667 | |
| -20°C | |
| Liquid |