missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Phospho-Jak1 (Tyr1034, Tyr1035) Recombinant Rabbit Monoclonal Antibody (Jak1Y10341035-F11), PE

Rabbit Recombinant Monoclonal Antibody
Brand: Invitrogen™ MA536891
This item is not returnable.
View return policy
Description
Phospho-Jak1 (Tyr1034, Tyr1035) Recombinant Monoclonal Antibody for Flow
Janus kinase 1 (JAK1), is a protein tyrosine kinase that is involved in the interferon-alpha/beta and -gamma signal transduction pathways. Immunological stimuli, such as interferons and cytokines, induce recruitment of Stat transcription factors to cytokine receptor-associated JAK1. JAK1 then phosphorylates proximal Stat factors, which subsequently dimerize, translocate to the nucleus and bind to cis elements upstream of target gene promoters to regulate transcription. Upon ligand binding, JAK1 undergoes tyrosine phosphorylation and catalytic activation in an interdependent manner. Phosphorylation of tyrosine residues at positions 1022 and 1023 is believed to function in the activation of catalytic events.
Specifications
| Phospho-Jak1 (Tyr1034, Tyr1035) | |
| Recombinant Monoclonal | |
| PE | |
| JAK1 | |
| AA960307; BAP004; C130039L05Rik; EC 2.7.10.2; Jak1; JAK-1; JAK1A; JAK1B; Janus kinase 1; Janus protein tyrosine kinase 1; JTK3; kinase Jak1; Tyrosine-protein kinase JAK1 | |
| Rabbit | |
| Protein A/G | |
| RUO | |
| 16451, 3716 | |
| 4°C | |
| Liquid |
| Flow Cytometry | |
| Jak1Y10341035-F11 | |
| PBS with 0.20% BSA and 0.09% sodium azide | |
| P23458, P52332 | |
| JAK1 | |
| A synthetic phospho-peptide corresponding to residues surrounding Tyr1034/1035 of human phospho-Jak1. | |
| 10 Tests | |
| Primary | |
| Human, Mouse | |
| Antibody | |
| IgG κ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction