missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Phospho-JMJD2B (Ser566) Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA572371
This item is not returnable.
View return policy
Description
Phospho-JMJD2B (Ser566) Polyclonal Antibody for DB
Histone demethylase that specifically demethylates 'Lys-9' of histone H3, thereby playing a role in histone code. Does not demethylate histone H3 'Lys-4', H3 'Lys-27', H3 'Lys-36' nor H4 'Lys-20'. Only able to demethylate trimethylated H3 'Lys-9', with a weaker activity than KDM4A, KDM4C and KDM4D. Demethylation of Lys residue generates formaldehyde and succinate. [UniProt].
Specifications
| Phospho-JMJD2B (Ser566) | |
| Polyclonal | |
| Unconjugated | |
| KDM4B | |
| 4732474L06Rik; JHDM3B; JmjC domain-containing histone demethylation protein 3B; Jmjd2b; jumonji domain containing 2B; jumonji domain-containing protein 2B; KDM4B; KIAA0876; lysine (K)-specific demethylase 4B; lysine demethylase 4B; lysine-specific demethylase 4B; mKIAA0876; TDRD14B; tudor domain containing 14B | |
| Rabbit | |
| Antigen affinity chromatography, Protein A | |
| RUO | |
| 23030 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Dot Blot | |
| 0.35 mg/mL | |
| PBS with 0.09% sodium azide; pH 7.4 | |
| O94953 | |
| KDM4B | |
| KLH conjugated synthetic phosphopeptide corresponding to amino acid residues surrounding S566 of human JMJD2B. | |
| 400 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction