missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Phospho-JNK1 (Thr183, Tyr185) Monoclonal Antibody (M268)
Mouse Monoclonal Antibody
Brand: Invitrogen™ MA547630
This item is not returnable.
View return policy
Description
This antibody detects 46 and 54 kDa* proteins corresponding to the apparent molecular mass of JNK1 and JNK2 on SDS-PAGE immunoblots of human A431 and HeLa cells, as well as rat PC12 cells treated with calyculin A.
MAPK8 (JNK1) is activated by the presence of misfolded proteins in the endoplasmic reticulum. It is also a crucial mediator of insulin resistance and obesity. It belongs to the MAPK superfamily of stress-activated protein kinases. MAPKs are Serine-threonine protein kinases that are activated in response to a variety of extracellular stimuli, and mediate signal transduction from the cell surface to the nucleus. JNK is involved in a wide variety of cellular processes such as proliferation, differentiation, transcription regulation and development. JNK pathways are activated by stress and inflammatory signals. JNK is expressed as ten different isoforms due to differential mRNA splicing. The predominant forms are JNK1 and JNK2.
Specifications
| Phospho-JNK1 (Thr183, Tyr185) | |
| Monoclonal | |
| 0.25 mg/mL | |
| PBS with 1mg/mL BSA, 50% glycerol and 0.05% sodium azide | |
| P45983, P49185, Q91Y86 | |
| MAPK8 | |
| Clone M268 was generated from a dual phosphorylated peptide corresponding to amino acid residues surrounding Thr-183 and Tyr-185 in human JNK1. This sequence has high homology to the conserved site in rat and mouse JNK1 and JNK2. | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG1 |
| Western Blot, Immunocytochemistry | |
| M268 | |
| Unconjugated | |
| MAPK8 | |
| AI849689; c-Jun N-terminal kinase 1; FLJ12099; FLJ33785; JNK; JNK1; JNK1 beta1 protein kinase; jnk1a-1; JNK1A2; JNK21B1/2; JNK3; JNK3A; JNK-46; JUN N-terminal kinase; MAP kinase 10; MAP kinase 8; MAPK 10; MAPK 8; MAPK10; Mapk8; mapk8 protein; mapk8b; MGC50974; mitogen activated protein kinase 8; mitogen-activated protein kinase 8; mitogen-activated protein kinase 8 isoform JNK1 alpha1; mitogen-activated protein kinase 8 isoform JNK1 beta2; mitogen-activated protein kinase 8b; MK10; OTTHUMP00000019552; p493F12; p54 gamma; p54bSAPK; Phospho-JNK; PRKM10; PRKM8; protein kinase JNK1; protein kinase mitogen-activated 8; SAPK gamma; SAPK1; SAPK1C; stress-activated protein kinase 1; stress-activated protein kinase 1c; Stress-activated protein kinase JNK1; zgc:112379 | |
| Mouse | |
| Protein A | |
| RUO | |
| 116554, 26419, 5599 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction