missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Phospho-MAP1B (Thr1265) Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA523014
This item is not returnable.
View return policy
Description
Recommended Blocking Buffer for ICC/IF and IHC: 5% (v/v) normal horse serum, 5% (v/v) normal goat serum, 0.2% (w/v) L-lysine, 0.2% (v/v) Triton X-100 in PBS, pH 7.2.
The coenzyme NAD and its derivatives are involved in hundreds of metabolic redox reactions and are utilized in protein ADP-ribosylation, histone deacetylation, and in some Ca(2+) signaling pathways. NMNAT.
Specifications
| Phospho-MAP1B (Thr1265) | |
| Polyclonal | |
| Unconjugated | |
| MAP1B | |
| A230055D22; AI843217; claustrin; DKFZp686E1099; DKFZp686F1345; FLJ38954; futsch; I79_010564; LC1; MAP1 light chain LC1; MAP1(X); MAP1.2; MAP1B; MAP-1B; MAP1B heavy chain; MAP5; microtubule associated protein 1B; microtubule associated protein 1B; microtubule-associated protein 1B; microtubule-associated protein 1B; Microtubule-associated protein 1B (MAP 1B) (MAP1.2) (MAP1(X)); microtubule-associated protein 5; Mtap1b; Mtap5; Mtap-5; Neuraxin; PPP1R102; protein phosphatase 1, regulatory subunit 102 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 17755, 29456, 396174, 4131, 779460 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Frozen), Western Blot, Immunocytochemistry | |
| 0.12 mg/mL | |
| PBS with 0.05% sodium azide | |
| P14873, P15205, P46821 | |
| MAP1B | |
| Phosphopeptide made to a C-terminal portion of rat MAP1B. | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat, Monkey, Chicken, Fish, Reptile, Frog | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction