missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Phospho-MAPRE3 (Ser162) Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA5143684
This item is not returnable.
View return policy
Description
The antibody detects a 32 kDa* protein corresponding to the apparent molecular mass of phosphorylated EB3 on SDS-PAGE immunoblots of mouse brain. This reactivity is significantly reduced after lambda phosphatase treatment.
EB3 is a 282 amino acid homologue of EB1, a protein known to bind APC (adenomatous polyposis coli). It has been shown to bind to tubulin in vitro and microtubules in vivo. Northern blotting shows expressions to be mainly in brain, and to a lesser degree in muscle. EB3 interacts with APCL, a homologue of the tumor suppressor APC, and this interaction may be involved in the stabilization or extension of microtubules.
Specifications
| Phospho-MAPRE3 (Ser162) | |
| Polyclonal | |
| Unconjugated | |
| MAPRE3 | |
| AI790651; APC binding protein; EB1 protein family member 3; EB2; EB3; EBF3; EBF3-S; End-binding protein 3; Mapre3; microtubule associated protein RP/EB family member 3; microtubule-associated protein RP/EB family member 3; microtubule-associated protein, RP/EB family, member 3; RP3 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 100732, 22924, 298848 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| Western Blot | |
| 0.25 mg/mL | |
| PBS with 1mg/mL BSA, 50% glycerol and 0.05% sodium azide | |
| Q5XIT1, Q6PER3, Q9UPY8 | |
| MAPRE3 | |
| Phospho-EB3 (Ser-162) synthetic peptide (coupled to KLH) corresponding to amino acid residues surrounding serine 162 in human EB3. This peptide sequence is highly conserved in mouse and rat EB3, and is not conserved in EB1 and EB2. | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction