missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Phospho-MAZ (Thr72) Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA5106007
This item is not returnable.
View return policy
Description
Antibody detects endogenous levels of MAZ only when phosphorylated at Thr72.
May function as a transcription factor with dual roles in transcription initiation and termination. Binds to two sites, ME1a1 and ME1a2, within the MYC promoter having greater affinity for the former. Also binds to multiple G/C-rich sites within the promoter of the Sp1 family of transcription factors. Regulates inflammation-induced expression of serum amyloid A proteins. [UniProt].
Specifications
| Phospho-MAZ (Thr72) | |
| Polyclonal | |
| Unconjugated | |
| MAZ | |
| LOW QUALITY PROTEIN: myc-associated zinc finger protein; Maz; MAZI; MYC associated zinc finger protein; myc-associated zinc finger protein; MYC-associated zinc finger protein (purine-binding transcription factor); PUR1; Pur-1; purine-binding transcription factor; SAF-1; SAF-2; SAF-3; serum amyloid A activating factor 1; serum amyloid A activating factor 2; serum amyloid A-activating factor-1; Transcription factor Zif87; ZF87; Zif87; Zinc finger protein 801; zinc-finger protein, 87 kilodaltons; ZNF801 | |
| Rabbit | |
| Sequential chromatography | |
| RUO | |
| 17188, 4150, 691842 | |
| -20°C | |
| Liquid |
| Western Blot | |
| 1 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
| P56270, P56671 | |
| MAZ | |
| A synthesized peptide derived from human MAZ(Accession P56270), corresponding to amino acid residues around phosphorylated Thr72. | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction