missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Phospho-MECP2 (Ser80) Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA5143716
This item is not returnable.
View return policy
Description
The antibody detects a 75 kDa* protein corresponding to the molecular mass of MeCP2 on SDS-PAGE immunoblots of human PC3 cells treated with calyculin A and in mouse brain tissue. These reactivities are not observed after lambda phosphatase treatment.
MECP2 belongs to a family of nuclear proteins (including MBD1, MBD2, MBD3 and MBD4) that have a methyl-CpG binding domain. MECP2 is capable of binding specifically to methylated DNA. DNA methylation is the major modification of eukaryotic genomes and plays an essential role in mammalian development. MECP2 can also repress transcription from methylated gene promoters. In contrast to other MBD family members, MECP2 is X-linked and subject to X inactivation. MECP2 is dispensible in stem cells, but is essential for embryonic development. MECP2 gene mutations are the cause of most cases of Rett syndrome, a progressive neurologic developmental disorder and one of the most common causes of mental retardation in females.
Specifications
| Phospho-MECP2 (Ser80) | |
| Polyclonal | |
| Unconjugated | |
| Mecp2 | |
| 1500041B07Rik; AUTSX3; D630021H01Rik; DKFZp686A24160; Mbd5; MECP2; MeCp-2 protein; methyl CpG binding protein 2; methyl-CpG binding protein 2; methyl-CpG-binding protein 2; MRX16; MRX79; MRXS13; MRXSL; PPMX; RS; RTS; RTT; testis tissue sperm-binding protein Li 41a; WBP10 | |
| Rabbit | |
| Affinity chromatography | |
| RUO | |
| 17257, 29386, 4204 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
| 0.5 mg/mL | |
| PBS with 1mg/mL BSA, 50% glycerol and 0.05% sodium azide | |
| P51608, Q00566, Q9Z2D6 | |
| Mecp2 | |
| phospho-MeCP2 (Ser-80) synthetic peptide (coupled to KLH) corresponding to amino acid residues surrounding serine 80 in mouse MeCP2. This peptide sequence is highly conserved in rat and human MeCP2, and has low homology to other nuclear proteins. | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction