Learn More
Phospho-MEK1 (Ser298) Mouse Monoclonal Antibody, Proteintech
Mouse Monoclonal Antibody
€ 135.00 - € 360.00
Specifications
| Antigen | MEK1 (Ser298) |
|---|---|
| Clone | 3F10G10 |
| Concentration | 1000 μg/mL |
| Applications | Western Blot, ELISA |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
30236910
|
Proteintech
68047-1-IG-100UL |
100 μL |
€ 360.00
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
30235170
|
Proteintech
68047-1-IG-20UL |
20 μL |
€ 135.00
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
The Phospho-MEK1 (Ser298) antibody from Proteintech is a mouse monoclonal antibody generated with peptide of human Phospho-MEK1 (Ser298). This antibody recognizes human antigen. The Phospho-MEK1 (Ser298) antibody has been validated for the following applications: WB, ELISA analysis.Specifications
| MEK1 (Ser298) | |
| 1000 μg/mL | |
| Monoclonal | |
| Liquid | |
| RUO | |
| 5604 | |
| Peptide | |
| Primary | |
| Store at -20°C. |
| 3F10G10 | |
| Western Blot, ELISA | |
| Unconjugated | |
| Mouse | |
| Human | |
| MAPKK1; MEK1; MKK1; PRKMK1 | |
| IgG1 | |
| Protein G purification | |
| Monoclonal Antibody |
Ser du en mulighed for forbedring?Del en indholdskorrektion
Korrektion af produktindhold
Dit input er vigtigt for os. Udfyld denne formular for at give feedback relateret til indholdet på dette produkt.
Produkttitel