missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Phospho-MKP-1 (Ser359) Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Invitrogen PA538677
This item is not returnable.
View return policy
Description
Dual specificity phosphatase that dephosphorylates MAP kinase MAPK1/ERK2 on both ′Thr-183′ and ′Tyr-185′, regulating its activity during the meiotic cell cycle.
Specifications
| Phospho-MKP-1 (Ser359) | |
| Polyclonal | |
| Unconjugated | |
| DUSP1 | |
| 3CH134, CL100, Dual specificity protein phosphatase 1, Dual specificity protein phosphatase hVH1, DUS1, DUSP1, EC 3.1.3.16, EC 3.1.3.48, MAP kinase phosphatase-1, MKP1, Protein-tyrosine phosphatase 3CH134, Protein-tyrosine phosphatase CL100, Protein-tyros | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 1843, 19252 | |
| -20°C | |
| Liquid |
| Immunohistochemistry (Paraffin) | |
| 1 mg/mL | |
| Dulbecco′s PBS with 150mM NaCl, 50% glycerol and 0.02% sodium azide; pH 7.4 | |
| P28562, P28563 | |
| DUSP1 | |
| A synthetic phosphopeptide derived from human MKP1 around the phosphorylation site of Ser359(L-Q-SP-P-I) | |
| 100 μg | |
| Primary | |
| Human, Mouse | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction