Learn More
Invitrogen™ Phospho-MLK3 (Ser674) Polyclonal Antibody
GREENER_CHOICE
Rabbit Polyclonal Antibody
Brand:  Invitrogen™ PA540303
Product Code. 15546991
  • € 520.00 / 100µL
Description
This antibody is predicted to react with human and rat based on sequence homology.
MAP3K11 (MLK3) is a serine/threonine kinase that activates both the p38 MAP kinase and c-Jun kinase pathways. This kinase contains a SH3 domain and a leucine zipper-basic motif. This kinase preferentially activates MAPK8/JNK kinase, and functions as a positive regulator of JNK signaling pathway. This kinase can directly phosphorylate, and activates IkappaB kinase alpha and beta, and is found to be involved in the transcription activity of NF-kappaB mediated by Rho family GTPases and CDC42.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Phospho-MLK3 (Ser674)
Polyclonal
Unconjugated
MAP3K11
2610017K16Rik; MAP3K11; MEKK11; MGC17114; mitogen activated protein kinase kinase kinase 11; mitogen-activated protein kinase kinase kinase 11; Mixed lineage kinase 3; Mlk3; MLK-3; protein-tyrosine kinase PTK1; PTK1; RHOE; SH3 domain-containing proline-rich kinase; SPRK; Src-homology 3 domain-containing proline-rich kinase
Rabbit
Antigen affinity chromatography
RUO
26403, 309168, 4296
-20°C, Avoid Freeze/Thaw Cycles
Liquid
ELISA, Western Blot
1 mg/mL
PBS with 50% glycerol, 0.5% BSA and 0.02% sodium azide; pH 7.4
Q16584, Q66HA1, Q80XI6
MAP3K11
Synthesized peptide derived from human MLK3 around the phosphorylation site of S674.
100 μL
Primary
Human, Mouse, Rat