missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Phospho-MNK1 (Thr250/255) Rabbit Recombinant Antibody, Proteintech
Rabbit Recombinant Antibody
€ 149.00 - € 439.00
Specifications
| Antigen | MNK1 (Thr250/255) |
|---|---|
| Clone | 4P9 |
| Concentration | 500 μg/mL |
| Applications | Western Blot, ELISA |
| Classification | Recombinant Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
30238108
|
Proteintech
81398-1-RR-20UL |
20 μL |
€ 149.00
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
30236705
|
Proteintech
81398-1-RR-100UL |
100 μL |
€ 439.00
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
The Phospho-MNK1 (Thr250/255) antibody from Proteintech is a rabbit recombinant antibody generated with peptide of human Phospho-MNK1 (Thr250/255). This antibody recognizes human antigen. The Phospho-MNK1 (Thr250/255) antibody has been validated for the following applications: WB, ELISA analysis.Specifications
| MNK1 (Thr250/255) | |
| 500 μg/mL | |
| Recombinant Monoclonal | |
| Liquid | |
| RUO | |
| 8569 | |
| Peptide | |
| Primary | |
| Store at -20°C. Stable for one year after shipment. |
| 4P9 | |
| Western Blot, ELISA | |
| Unconjugated | |
| Rabbit | |
| Human | |
| MNK1 | |
| IgG | |
| Protein A purification | |
| Recombinant Antibody |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title