missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Phospho-MST1/MST2 (Thr183) Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Invitrogen PA538372
This item is not returnable.
View return policy
Description
The mammalian Ste20-like kinases (MST kinases), also known as Krs proteins, are members of the GCK subfamily. Ksr-1 (MST-2) and Ksr-2 (MST-1) are both direct substrates of caspase-3 that accelerate caspase-3 activation. MST-3 is ubiquitously expressed in mammalian tissue and can phosphorylate exogenous substrates as well as itself. MST-4 is highly expressed in placenta, thymus, and peripheral blood leukocytes, and it specifically activates ERK.
Specifications
| Phospho-MST1/MST2 (Thr183) | |
| Polyclonal | |
| Unconjugated | |
| STK3 | |
| Serine/threonine-protein kinase 4, Mammalian STE20-like protein kinase 1, MST-1, STE20-like kinase MST1, Serine/threonine-protein kinase Krs-2, MST1/C STK4, KRS2, MST1, Serine/threonine-protein kinase 3, Mammalian STE20-like protein kinase 2, MST-2, STE20-like kinase MST2, Serine/threonine-protein kinase Krs-1, MST2/C, STK3, KRS1, MST2 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 56274, 58231, 6788, 6789 | |
| -20°C | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot | |
| 1 mg/mL | |
| Dulbecco′s PBS with 150mM NaCl, 50% glycerol and 0.02% sodium azide; pH 7.4 | |
| Q13043, Q13188, Q9JI10, Q9JI11 | |
| STK3, STK4 | |
| A synthetic phosphopeptide derived from human Mst1/2 around the phosphorylation site of Thr183(R-N-TP-V-I) | |
| 100 μg | |
| Primary | |
| Human, Mouse | |
| Antibody | |
| IgG |
Product Content Correction
Bitte geben Sie uns Ihr Feedback zu den Produktinhalten, indem Sie das folgende Formular ausfüllen.
Name des Produkts
Haben Sie Verbesserungsvorschläge?Übermitteln Sie eine inhaltliche Korrektur