missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Phospho-MyD88 (Tyr257) Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA5106188
This item is not returnable.
View return policy
Description
Antibody detects endogenous levels of MyD88 only when phosphorylated at Tyr257.
Myeloid differentiation Marker 88 (MyD88) is an adaptor molecule for the Toll/IL-1R and is involved in the inflammatory response induced by IL-1, IL-18 and LPS. MyD88 recruits IRAK to IL-1 receptor complex in response to IL-1. This pathway further leads to activation of NF-kB. MyD88 has a wide tissue distribution.
Specifications
| Phospho-MyD88 (Tyr257) | |
| Polyclonal | |
| Unconjugated | |
| MYD88 | |
| mutant myeloid differentiation primary response 88; Myd88; MYD88D; myeloid differentiation primary response 88; myeloid differentiation primary response gene (88); myeloid differentiation primary response gene 88; myeloid differentiation primary response protein MyD88 | |
| Rabbit | |
| sequential chromatography | |
| RUO | |
| 17874, 301059, 4615 | |
| -20°C | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot | |
| 1 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
| P22366, Q6Y1S1, Q99836 | |
| MYD88 | |
| A synthesized peptide derived from human MYD88(Accession Q99836), corresponding to amino acid residues around phosphorylated Tyr257. | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction