missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Phospho-MYH9 (Ser1916) Polyclonal Antibody

Phospho-MYH9 (Ser1916) Polyclonal Antibody
Brand: Invitrogen PA5143719
This item is not returnable.
View return policy
Description
Phospho-MYH9 (Ser1916) Polyclonal Antibody for Western Blot
Specifications
| Phospho-MYH9 (Ser1916) | |
| Polyclonal | |
| Unconjugated | |
| MYH9 | |
| BDPLT6; Cellular myosin heavy chain, type A; DFNA17; epsts; fi22c04; fj85e11; flectin; Fltn; FTNS; KLG/PTK7; LOC100911597; LOW QUALITY PROTEIN: myosin-9; MGC104539; MHA; Myh9; myh9.L; myh9a; myh9l2; Myhn1; Myhn-1; myosin; Myosin Heavy Chain 2A; myosin heavy chain 9; myosin heavy chain IX; myosin heavy chain, non-muscle IIa; myosin IIA; myosin, heavy chain 9, non-muscle; myosin, heavy chain 9, non-muscle L homeolog; myosin, heavy chain 9, non-muscle, like-2; myosin, heavy chain 9a, non-muscle; myosin, heavy polypeptide 9, non-muscle; myosin, heavy polypeptide 9a, non-muscle; myosin-2a; myosin9; Myosin-9; myosin-9; myosin-9a; myosin-9-like; NMHC II-A; NMHC-II A; NMHCIIA; NMHC-II-A; NMMHC IIA; NMMHC II-a; nmmhca; NMMHC-A; NMMHC-IIA; non-muscle myosin heavy chain 9; non-muscle myosin heavy chain A; non-muscle myosin heavy chain II A; non-muscle myosin heavy chain IIa; nonmuscle myosin heavy chain II-A; non-muscle myosin heavy polypeptide 9; nonmuscle myosin II heavy chain A; TU72.6; Unknown (protein for IMAGE:7151033); Unknown (protein for IMAGE:7177375); Unknown (protein for IMAGE:7268224); wu:fi22c04; wu:fi22c04 protein; wu:fj85e11; XELAEV_18023533mg; zgc:162029; zgc:66164; zNMHC-IIA | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 100911597, 17886, 4627 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| Western Blot | |
| ~1.0 mg/mL | |
| PBS with 1mg/mL BSA, 50% glycerol and 0.05% sodium azide | |
| P35579, Q62812, Q8VDD5 | |
| MYH9 | |
| Phospho-Myosin IIA Heavy Chain (Ser-1916) synthetic peptide (coupled to KLH) corresponding to amino acid residues surrounding Ser-1916 in human myosin IIA heavy chain. This peptide sequence is highly conserved in rat and mouse myosin IIA heavy chain. | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction