missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Phospho-NF2 (Ser518) Polyclonal Antibody
Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA128383
This item is not returnable.
View return policy
Description
PA1-28383 detects NF2 from mouse, human samples.
NF2 encodes a protein, Merlin, that is similar to some members of the ERM family of proteins that are thought to link cytoskeletal components with proteins in the cell membrane. Merlin has been shown to interact with cell-surface proteins, proteins involved in cytoskeletal dynamics and proteins involved in regulating ion transport. This gene is expressed at high levels during embryonic development; in adults, significant expression is found in Schwann cells, meningeal cells, lens and nerve. Mutations in this gene are associated with neurofibromatosis type II which is characterized by nervous system and skin tumors and ocular abnormalities. Two predominant isoforms and a number of minor isoforms are produced by alternatively spliced transcripts.
Specifications
| Phospho-NF2 (Ser518) | |
| Polyclonal | |
| Unconjugated | |
| Nf2 | |
| ACN; BANF; Merlin; MGC123124; moesin-ezrin-radixin like; moesin-ezrin-radixin-like protein; moesin-ezrin-radizin-like protein; neurofibromatosis 2; neurofibromin 2; neurofibromin 2 (bilateral acoustic neuroma); neurofibromin 2 (merlin); neurofibromin 2b (merlin); neurofibromin-2; Nf2; Nf-2; NF2a; nf2b; OTTHUMP00000199607; SCH; Schwannomerlin; schwannomin | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 18016, 4771 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| ELISA, Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
| 1 mg/mL | |
| PBS with 0.15M NaCl and 0.01% sodium azide; pH 7.2 | |
| P35240, P46662 | |
| Nf2 | |
| Human NF2 (Merlin) phospho-peptide corresponding to a region of the human protein surrounding S518 and conjugated to KLH. | |
| 50 μg | |
| Primary | |
| Human, Mouse | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction