missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Phospho-NHE3 (Ser552) Monoclonal Antibody (14D5), Biotin

Mouse Monoclonal Antibody
Brand: Invitrogen™ MA516092
This item is not returnable.
View return policy
Description
Phospho-NHE3 (Ser552) Monoclonal Antibody for Western Blot, IHC, ICC/IF, Flow, ELISA, IP
NHE3 (also known as SLC9A3 - Sodium/hydrogen exchanger 3) is a member of the sodium/hydrogen exchanger family that plays a crucial role in acid-base and volume homeostasis by mediating the majority of sodium and bicarbonate reabsorption in the proximal tubule of the kidney. Mutations in the gene can lead to Diarrhea 8.
Specifications
| Phospho-NHE3 (Ser552) | |
| Monoclonal | |
| 1.55 mg/mL | |
| PBS with 0.05% sodium azide | |
| G3X939, P26432, P26433, P48764 | |
| SLC9A3 | |
| Synthetic peptide made to a region surrounding the Serine 552 portion of rat NHE3. | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat, Rabbit, Monkey, Pig, Marsupial | |
| Antibody | |
| IgG2a |
| ELISA, Flow Cytometry, Immunohistochemistry, Immunoprecipitation, Western Blot, Immunocytochemistry | |
| 14D5 | |
| Biotin | |
| SLC9A3 | |
| 9030624O13Rik; AI930210; DIAR8; MGC126718; MGC126720; Na(+)/H(+) exchanger 3; Na/H exchanger 3; Na+/H+ exchanger homolog; Na+/H+ exchanger isoform 3; membrane ion exchange protein; Nhe3; NHE-3; NHE-3 antibody; plasma membrane ion transport protein; putative sodium hydrogen exchanger subtype 3; Slc9a3; SLC9A3 antibody; sodium proton exchanger; sodium/hydrogen exchanger; sodium/hydrogen exchanger 3; sodium/hydrogen exchanger isoform 3; Solute carrier family 9 (sodium/hydrogen exchanger 3), antiporter 3, Na+/H+ (amiloride insensitive); solute carrier family 9 (sodium/hydrogen exchanger), isoform 3; solute carrier family 9 (sodium/hydrogen exchanger), member 3; solute carrier family 9 member 3; solute carrier family 9 member A3; solute carrier family 9, member 3; solute carrier family 9, subfamily A (NHE3, cation proton antiporter 3), member 3 | |
| Mouse | |
| Protein G | |
| RUO | |
| 100009430, 105243, 24784, 397358, 6550 | |
| 4°C, store in dark | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction