missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Phospho-NMDAR2A (Tyr943) Polyclonal Antibody
Description
Antibody detects endogenous levels of NMDAR2A only when phosphorylated at Tyr943.
N-methyl-D-aspartate (NMDA) receptors are a class of ionotropic glutamate-gated ion channels. These receptors have been shown to be involved in long-term potentiation, an activity-dependent increase in the efficiency of synaptic transmission thought to underlie certain kinds of memory and learning. NMDA receptor channels are heteromers composed of the key receptor subunit NMDAR1 (GRIN1) and 1 or more of the 4 NMDAR2 subunits: NMDAR2A (GRIN2A), NMDAR2B (GRIN2B), NMDAR2C (GRIN2C) and NMDAR2D (GRIN2D). Alternatively spliced transcript variants encoding different isoforms have been found for this gene.
Specifications
Specifications
| Antigen | Phospho-NMDAR2A (Tyr943) |
| Applications | Immunohistochemistry (Paraffin), Western Blot |
| Classification | Polyclonal |
| Concentration | 1 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 |
| Gene | Grin2a |
| Gene Accession No. | P35436, Q00959, Q12879 |
| Gene Alias | EPND; FESD; GluN2A; GluRepsilon1; glutamate [NMDA] receptor subunit epsilon-1; glutamate ionotropic receptor NMDA type subunit 2A; glutamate receptor; glutamate receptor ionotropic, NMDA 2A; glutamate receptor, ionotropic, NMDA2A (epsilon 1); glutamate receptor, ionotropic, N-methyl D-aspartate 2A; Grin2a; hNR2A; LKS; NMDA Receptor 2A; NMDAR 2A; NMDAR2A; N-methyl D-aspartate receptor subtype 2A; N-methyl-D-aspartate receptor channel, subunit epsilon-1; N-methyl-D-aspartate receptor subunit 2A; NR2A; OTTHUMP00000160135; OTTHUMP00000174531 |
| Gene Symbols | Grin2a |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?