missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Phospho-Nucleolin (Thr76) Recombinant Rabbit Monoclonal Antibody (5V6R7)

Product Code. 30132371
Change view
Click to view available options
Quantity:
100 μL
Unit Size:
100µL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30132371 100 μL 100µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30132371 Supplier Invitrogen™ Supplier No. MA555195

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Recombinant Monoclonal Antibody

Phospho-Nucleolin (Thr76) Recombinant Monoclonal Antibody for IHC (P), ELISA

Nucleolin (NCL), a eukaryotic nucleolar phosphoprotein, is involved in the synthesis and maturation of ribosomes. It is located mainly in dense fibrillar regions of the nucleolus. Human NCL gene consists of 14 exons with 13 introns and spans approximately 11kb. The intron 11 of the NCL gene encodes a small nucleolar RNA, termed U20.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Phospho-Nucleolin (Thr76)
Applications ELISA, Immunohistochemistry (Paraffin)
Classification Recombinant Monoclonal
Clone 5V6R7
Concentration 0.75 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.05% BSA, 50% glycerol and 0.09% sodium azide; pH 7.3
Gene NCL
Gene Accession No. P19338
Gene Alias B530004O11Rik; C23; D0Nds28; D1Nds28; FLJ45706; FLJ59041; Ncl; Nuc; Nucl; nucl antibody; Nucleolin; nucleolin-like protein; Protein C23
Gene Symbols NCL
Host Species Rabbit
Immunogen Synthetic Peptide.
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 4691
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.