missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Phospho-p130Cas (Tyr410) Recombinant Rabbit Monoclonal Antibody (23GB3870), Invitrogen™

Rabbit Recombinant Monoclonal Antibody
€ 207.00 - € 562.00
Specifications
| Antigen | Phospho-p130Cas (Tyr410) |
|---|---|
| Clone | 23GB3870 |
| Applications | Flow Cytometry, Western Blot, Immunocytochemistry |
| Classification | Recombinant Monoclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
30089951
![]() |
Invitrogen™
MA553656 |
100 μL |
€ 562.00
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
30088033
|
Invitrogen™
MA553657 |
20 μL |
€ 207.00
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Phospho-p130Cas (Tyr410) Recombinant Monoclonal antibody specifically detects Phospho-p130Cas (Tyr410) in Human, Mouse, Rat samples. It is validated for Flow Cytometry, Western BlotSpecifications
| Phospho-p130Cas (Tyr410) | |
| Flow Cytometry, Western Blot, Immunocytochemistry | |
| Unconjugated | |
| Rabbit | |
| Human, Mouse, Rat | |
| P56945, Q61140, Q63767 | |
| 12927, 25414, 9564 | |
| IgG | |
| Affinity chromatography | |
| Antibody |
| 23GB3870 | |
| Recombinant Monoclonal | |
| Liquid | |
| RUO | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
| AI385681; BCA1; Bcar1; BCAR1 scaffold protein, Cas family member; BCAR1, Cas family scaffold protein; BCAR1, Cas family scaffolding protein; breast cancer anti-estrogen resistance 1; breast cancer anti-estrogen resistance protein 1; Cas; Cas scaffolding protein family member 1; CAS1; CASS1; CRKAS; CRK-associated substrate; Crk-associated substrate p130Cas; OTTHUMP00000174941; p130 Cas; P130CAS; p130Cas (pTyr410); p130Cas (pY410); p130Cas phospho; phospho p130Cas; v-crk-associated tyrosine kinase substrate | |
| A synthetic peptide of human Phospho-BCAR1 (Tyr410) (403-417AA). | |
| Primary | |
| -20°C | |
| BCAR1 |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title