Learn More
Invitrogen™ Phospho-p130Cas (Tyr751) Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA5143733
Description
This antibody was cross-adsorbed to phospho-tyrosine coupled to agarose and to phospho-p130 Cas (Tyr-762) peptide before affinity purification using phospho-p130 Cas (Tyr-751) peptide (without carrier). The antibody detects a 130 kDa* protein corresponding to the molecular mass of phosphorylated p130 Cas on SDS-PAGE immunoblots of pervanadate treated human endothelial and A431 cells, but is not observed in control cells.
The p130Cas (Cas for Crk-associated substrate) is a common cellular target of phosphorylation signal via v-Crk and v-Src oncoproteins. p130Cas has a unique structure that contains a Src homology (SH)-3 domain followed by multiple YXXP motifs and a proline-rich regionp130Cas is implicated in a variety of biological processes including cell adhesion, cell migration, growth factor stimulation, cytokine receptor engagement and bacterial infection. Physiological functions of p130Cas include cardiovascular development, actin filament assembly and Src-induced cell transformation.
Specifications
| Phospho-p130Cas (Tyr751) | |
| Polyclonal | |
| Unconjugated | |
| BCAR1 | |
| AI385681; BCA1; Bcar1; BCAR1 scaffold protein, Cas family member; BCAR1, Cas family scaffold protein; BCAR1, Cas family scaffolding protein; breast cancer anti-estrogen resistance 1; breast cancer anti-estrogen resistance protein 1; Cas; Cas scaffolding protein family member 1; CAS1; CASS1; CRKAS; CRK-associated substrate; Crk-associated substrate p130Cas; OTTHUMP00000174941; p130 Cas; P130CAS; p130Cas (pTyr410); p130Cas (pY410); p130Cas phospho; phospho p130Cas; v-crk-associated tyrosine kinase substrate | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 12927, 25414, 9564 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| Western Blot | |
| 0.5 mg/mL | |
| PBS with 1mg/mL BSA, 50% glycerol and 0.05% sodium azide | |
| P56945, Q61140, Q63767 | |
| BCAR1 | |
| Phospho-p130 Cas (Tyr-751) synthetic peptide (coupled to KLH) corresponding to amino acid residues around tyrosine 751 in rat p130 Cas. This rat sequence is the same as Tyr-653 in human p130 Cas and Tyr-657 in mouse p130 Cas. | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.