Learn More
Phospho-p27 Kip1 (Ser178) Polyclonal Antibody
GREENER_CHOICE
Phospho-p27 Kip1 (Ser178) Polyclonal Antibody
Brand:  Invitrogen PA599567
Product Code. 16663815
  • € 494.00 / 100µL
Description
Phospho-p27 Kip1 (Ser178) Polyclonal Antibody for Western Blot, ICC/IF, IHC (P)
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Phospho-p27 Kip1 (Ser178)
Polyclonal
Unconjugated
Cdkn1b
AA408329; AI843786; Cdki1b; CDKN1B; CDKN4; CDN1B; cyclin dependent kinase inhibitor 1B; cyclin-dependent kinase inhibitor 1B; Cyclin-dependent kinase inhibitor 1B (p27 Kip1); Cyclin-dependent kinase inhibitor 1B (p27, Kip1); cyclin-dependent kinase inhibitor p27; Cyclin-dependent kinase inhibitor p27 Kip1; cyclin-dependent kinase inhibitor p27/Kip1; KIP1; MEN1B; MEN4; p27; P27 CDKN1B; p27(KIP1); p27Kip1
Rabbit
Sequential Chromatography
RUO
1027, 12576, 83571
-20°C
Liquid
Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
1 mg/mL
PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
P46414, P46527
Cdkn1b
A synthesized peptide derived from human CDKN1B(Accession P46527), corresponding to amino acid residues around phosphorylated Ser178.