missing translation for 'onlineSavingsMsg'
Learn More

Phospho-p38 alpha (T180/Y182) DuoSet IC ELISA

Product Code. 30021632
Click to view available options
Format:
15 96-well Plates
2 96-well Plates
5 96-well Plates
Unit Size:
1 set
This item is not returnable. View return policy

Product Code. 30021632

Brand: R&D Systems DYC869B2

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Phospho-p38 alpha (T180/Y182) DuoSet IC ELISA from the most referenced ELISA manufacturer. Highly validated for accurate quantitation and long-term reproducibility.

Complete ELISpot kits are highly sensitive, microplate-based assays for the detection of cytokine secreting cells. Kits are available for detection and enumeration of a single analyte or two analytes simultaneously.

The Phospho-p38 alpha (T180/Y182) DuoSet IC ELISA is a Solid Phase Sandwich ELISA that quantifies phosphorylated human, mouse, rat p38 alpha.
TRUSTED_SUSTAINABILITY

Specifications

Product Type ELISA Development Kit for Phosphorylated Intracellular Proteins
Quantity 1 Kit (for 2 Plates)
Species Human, Mouse, Rat
Conjugate Biotin
Format 2 96-well Plates
For Use With (Application) ELISA
Gene ID (Entrez) 1432
Content And Storage Store the unopened product at 2°C to 8°C. Do not use past expiration date.
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.