missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Phospho-p38 alpha (T180/Y182) DuoSet IC ELISA
Click to view available options
Format:
15 96-well Plates
2 96-well Plates
5 96-well Plates
Unit Size:
1 set
Description
Complete ELISpot kits are highly sensitive, microplate-based assays for the detection of cytokine secreting cells. Kits are available for detection and enumeration of a single analyte or two analytes simultaneously.
The Phospho-p38 alpha (T180/Y182) DuoSet IC ELISA is a Solid Phase Sandwich ELISA that quantifies phosphorylated human, mouse, rat p38 alpha.
Specifications
Specifications
| Product Type | ELISA Development Kit for Phosphorylated Intracellular Proteins |
| Quantity | 1 Pk. (for 15 Plates) |
| Species | Human, Mouse, Rat |
| Conjugate | Biotin |
| Format | 15 96-well Plates |
| For Use With (Application) | ELISA |
| Gene ID (Entrez) | 1432 |
| Content And Storage | Store the unopened product at 2°C to 8°C. Do not use past expiration date. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction