Learn More
Phospho-P53 (Ser15) Mouse, anti-Human, Unconjugated, Clone: 1H6G1, Proteintech
Mouse Monoclonal Antibody
Brand:  Proteintech 67826-1-IG-100UL
 View more versions of this product
Product Code. 17241476
  • € 360.00 / 100µL
Description
TP53, also known as P53 and NY-CO-13, belongs to the p53 family and has 9 isoforms. In SDS-Page, the observed molecular weight is about 53 kDa. TP53 acts as a tumor suppressor in many tumor types, including growth arrest or apoptosis depending on the physiological circumstances and cell types. It is involved in cell cycle regulation as a trans-activator that acts to negatively regulate cell division by controlling a set of genes required for this process. TP53 Localizes in the nucleus in most cells but found in the cytoplasm in some cells. (PMID: 26166714; PMID: 25225161)
Specifications
Specifications
Phospho-P53 (Ser15)
Monoclonal
1000 μg/mL
Mouse
Primary
Human
Western Blot, ELISA
1H6G1
Unconjugated
100 μL
7157