missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Phospho-p73 (Ser388) Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA5106198
This item is not returnable.
View return policy
Description
Antibody detects endogenous levels of p73 only when phosphorylated at Ser388.
P73 (TP73) participates in the apoptotic response to DNA damage. Isoforms containing the transactivation domain are pro-apoptotic, isoforms lacking the domain are anti-apoptotic and block the function of p53 and transactivating p73 isoforms. May be a tumor suppressor protein.
Specifications
| Phospho-p73 (Ser388) | |
| Polyclonal | |
| Unconjugated | |
| TP73 | |
| deltaNp73; OTTHUMP00000003479; OTTHUMP00000003480; p53-like transcription factor; p53-related protein; P73; RP23-254N4.10; TAp73; Tp73; transformation related protein 73; Trp73; Tumor protein p73 | |
| Rabbit | |
| Sequential chromatography | |
| RUO | |
| 22062, 7161 | |
| -20°C | |
| Liquid |
| Western Blot | |
| 1 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
| O15350, Q9JJP2 | |
| TP73, Trp73 | |
| A synthesized peptide derived from human TP73(Accession O15350), corresponding to amino acid residues around phosphorylated Ser388. | |
| 100 μL | |
| Primary | |
| Human, Mouse | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction