missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Phospho-PAX7 (Ser203) Polyclonal Antibody
GREENER_CHOICE

Product Code. 16641805
Change view
Click to view available options
Quantity:
100 μL
Unit Size:
100µL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16641805 100 μL 100µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 16641805 Supplier Invitrogen™ Supplier No. PA5105359

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

Antibody detects endogenous levels of PAX7 only when phosphorylated at Ser203.

This gene is a member of the paired box (PAX) family of transcription factors. Members of this gene family typically contain a paired box domain, an octapeptide, and a paired-type homeodomain. These genes play critical roles during fetal development and cancer growth. The specific function of the paired box 7 gene is unknown but speculated to involve tumor suppression since fusion of this gene with a forkhead domain family member has been associated with alveolar rhabdomyosarcoma. Three transcript variants encoding different isoforms have been found for this gene.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Phospho-PAX7 (Ser203)
Applications Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
Gene PAX7
Gene Accession No. P23759, P47239
Gene Alias FLJ37460; HUP1; paired box 7; paired box gene 7; paired box homeotic gene 7; paired box protein Pax-7; paired box transcription factor PAX7; paired domain gene 7; PAX7; Pax-7; PAX7 transcriptional factor; PAX7A; PAX7B; RGD1564360; RMS2
Gene Symbols PAX7
Host Species Rabbit
Immunogen A synthesized peptide derived from human PAX7(Accession P23759), corresponding to amino acid residues around phosphorylated Ser203.
Purification Method sequential chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 18509, 5081
Target Species Human, Mouse
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.