Learn More
Invitrogen™ Phospho-PDGFRB (Tyr1021) Recombinant Superclonal™ Antibody (6HCLC)
Rabbit Recombinant Superclonal Antibody
Brand: Invitrogen™ 711359
Description
This antibody is predicted to react with Monkey, Bovine and Sheep Recombinant rabbit Superclonal™ antibodies are unique offerings from Thermo Fisher Scientific. They are comprised of a selection of multiple different recombinant monoclonal antibodies, providing the best of both worlds - the sensitivity of polyclonal antibodies with the specificity of monoclonal antibodies - all delivered with the consistency only found in a recombinant antibody. While functionally the same as a polyclonal antibody - recognizing multiple epitope sites on the target and producing higher detection sensitivity for low abundance targets - a recombinant rabbit Superclonal™ antibody has a known mixture of light and heavy chains. The exact population can be produced in every lot, circumventing the biological variability typically associated with polyclonal antibody production. Note: Formerly called Recombinant polyclonal antibody, this product is now rebranded as Recombinant Superclonal™ antibody. The physical product and the performance remain unchanged.
PDGFRB is a receptor tyrosine kinase and has been implicated in atherogenesis and in cell transformation by the SIS oncogene. These growth factors are mitogens for cells of mesenchymal origin. The identity of the growth factor bound to a receptor monomer determines whether the functional receptor is a homodimer or a heterodimer, composed of both platelet-derived growth factor receptor alpha and beta polypeptides. The gene is flanked on chromosome 5 by the genes for granulocyte-macrophage colony-stimulating factor and macrophage-colony stimulating factor receptor; all three genes may be implicated in the 5-q syndrome. A translocation between chromosomes 5 and 12, that fuses PDGFRB (CD140b) to that of the translocation, ETV6, leukemia gene, results in chronic myeloproliferative disorder with eosinophilia.
Specifications
| Phospho-PDGFRB (Tyr1021) | |
| Recombinant Superclonal | |
| 0.5 mg/mL | |
| PBS with 0.09% sodium azide; pH 7.4 | |
| P05622, P09619 | |
| Pdgfrb | |
| Peptide corresponding to Human PDGFRB (aa 1016-1024). | |
| 100 μg | |
| Primary | |
| Human, Mouse | |
| Antibody | |
| IgG |
| Immunocytochemistry | |
| 6HCLC | |
| Unconjugated | |
| Pdgfrb | |
| AI528809; Beta platelet-derived growth factor receptor; beta-type platelet-derived growth factor receptor; CD140 antigen-like family member B; CD140B; CD140b antigen; IBGC4; IMF1; JTK12; KOGS; PDGF beta chain; PDGF Receptor beta; Pdgfr; Pdgfr1; PDGFR-1; PDGFRB; PDGF-R-beta; PDGFR-beta; PENTT; platelet derived growth factor receptor beta; platelet derived growth factor receptor, beta polypeptide; platelet-derived growth factor receptor 1; Platelet-derived growth factor receptor beta; platelet-derived growth factor receptor beta variant 1; Platelet-derived growth factor receptor, beta; platelet-derived growth factor receptor, beta polypeptide | |
| Rabbit | |
| Protein A | |
| RUO | |
| 18596, 5159 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.